Product Details

SNP ID
rs144510037
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:46469437 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAGCCCCGGGACATTCTGCATTC[A/G]ATATTCTATGGTCTTGCTTCTTTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD26P1 PubMed Links

Gene Details

Gene
ANKRD26P1
Gene Name
ankyrin repeat domain 26 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details