Product Details
- SNP ID
-
rs144616330
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:67727336 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAATAGATTCACTCACCTAAAATGG[C/T]GCTGTTGAGGGCAGCACACACAGGT
- Phenotype
-
MIM: 614031
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GFOD2
PubMed Links
Gene Details
- Gene
- GFOD2
- Gene Name
- glucose-fructose oxidoreductase domain containing 2
There are no transcripts associated with this gene.
- Gene
- RANBP10
- Gene Name
- RAN binding protein 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001320238.1 |
1353 |
Missense Mutation |
ACC,GCC |
T488A |
NP_001307167.1 |
NM_001320239.1 |
1353 |
Missense Mutation |
ACC,GCC |
T549A |
NP_001307168.1 |
NM_001320240.1 |
1353 |
Missense Mutation |
ACC,GCC |
T346A |
NP_001307169.1 |
NM_020850.2 |
1353 |
Missense Mutation |
ACC,GCC |
T575A |
NP_065901.1 |
XM_005256070.2 |
1353 |
Missense Mutation |
ACC,GCC |
T605A |
XP_005256127.1 |
XM_005256072.2 |
1353 |
Missense Mutation |
ACC,GCC |
T473A |
XP_005256129.1 |
XM_006721238.3 |
1353 |
Missense Mutation |
ACC,GCC |
T449A |
XP_006721301.1 |
XM_011523245.2 |
1353 |
Missense Mutation |
ACC,GCC |
T346A |
XP_011521547.1 |
XM_011523247.2 |
1353 |
Missense Mutation |
ACC,GCC |
T376A |
XP_011521549.1 |
XM_017023507.1 |
1353 |
Missense Mutation |
ACC,GCC |
T391A |
XP_016878996.1 |
XM_017023508.1 |
1353 |
Missense Mutation |
ACC,GCC |
T376A |
XP_016878997.1 |
XM_017023509.1 |
1353 |
Missense Mutation |
ACC,GCC |
T346A |
XP_016878998.1 |
View Full Product Details