Product Details
- SNP ID
-
rs144919029
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:57760762 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAGGGTCTTGCTGTCACCACTAAG[C/T]GAATCCTGCAGCAGGTAGGTGAGCT
- Phenotype
-
MIM: 602703
MIM: 604535
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KATNB1
PubMed Links
Gene Details
- Gene
- KATNB1
- Gene Name
- katanin regulatory subunit B1
There are no transcripts associated with this gene.
- Gene
- KIFC3
- Gene Name
- kinesin family member C3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001130099.1 |
2949 |
Silent Mutation |
TCA,TCG |
S593S |
NP_001123571.1 |
NM_001130100.1 |
2949 |
Silent Mutation |
TCA,TCG |
S732S |
NP_001123572.1 |
NM_001318710.1 |
2949 |
Silent Mutation |
TCA,TCG |
S754S |
NP_001305639.1 |
NM_001318711.1 |
2949 |
Silent Mutation |
TCA,TCG |
S674S |
NP_001305640.1 |
NM_001318712.1 |
2949 |
Silent Mutation |
TCA,TCG |
S630S |
NP_001305641.1 |
NM_001318713.1 |
2949 |
Silent Mutation |
TCA,TCG |
S590S |
NP_001305642.1 |
NM_001318714.1 |
2949 |
Silent Mutation |
TCA,TCG |
S593S |
NP_001305643.1 |
NM_001318715.1 |
2949 |
Silent Mutation |
TCA,TCG |
S593S |
NP_001305644.1 |
NM_005550.3 |
2949 |
Silent Mutation |
TCA,TCG |
S732S |
NP_005541.3 |
XM_005255937.1 |
2949 |
Silent Mutation |
TCA,TCG |
S895S |
XP_005255994.1 |
XM_005255944.1 |
2949 |
Silent Mutation |
TCA,TCG |
S674S |
XP_005256001.1 |
XM_006721188.1 |
2949 |
Silent Mutation |
TCA,TCG |
S921S |
XP_006721251.1 |
XM_011523075.1 |
2949 |
Silent Mutation |
TCA,TCG |
S781S |
XP_011521377.1 |
XM_011523076.1 |
2949 |
Silent Mutation |
TCA,TCG |
S781S |
XP_011521378.1 |
XM_011523077.1 |
2949 |
Silent Mutation |
TCA,TCG |
S760S |
XP_011521379.1 |
XM_011523078.1 |
2949 |
Silent Mutation |
TCA,TCG |
S593S |
XP_011521380.1 |
XM_011523079.1 |
2949 |
Silent Mutation |
TCA,TCG |
S593S |
XP_011521381.1 |
XM_017023219.1 |
2949 |
Silent Mutation |
TCA,TCG |
S978S |
XP_016878708.1 |
XM_017023220.1 |
2949 |
Silent Mutation |
TCA,TCG |
S956S |
XP_016878709.1 |
XM_017023221.1 |
2949 |
Silent Mutation |
TCA,TCG |
S895S |
XP_016878710.1 |
XM_017023222.1 |
2949 |
Silent Mutation |
TCA,TCG |
S811S |
XP_016878711.1 |
XM_017023223.1 |
2949 |
Silent Mutation |
TCA,TCG |
S754S |
XP_016878712.1 |
XM_017023224.1 |
2949 |
Silent Mutation |
TCA,TCG |
S811S |
XP_016878713.1 |
XM_017023225.1 |
2949 |
Silent Mutation |
TCA,TCG |
S675S |
XP_016878714.1 |
XM_017023226.1 |
2949 |
Silent Mutation |
TCA,TCG |
S674S |
XP_016878715.1 |
XM_017023227.1 |
2949 |
Silent Mutation |
TCA,TCG |
S674S |
XP_016878716.1 |
View Full Product Details