Product Details

SNP ID
rs144972780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31061788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCGAGTGCACCCGGCTATGCTT[C/T]GTGAGGCTGGCACGCTCGGCGAAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF668 PubMed Links

Gene Details

Gene
ZNF668
Gene Name
zinc finger protein 668
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172668.1 1451 Silent Mutation ACA,ACG T380T NP_001166139.1
NM_001172669.1 1451 Silent Mutation ACA,ACG T403T NP_001166140.1
NM_001172670.1 1451 Silent Mutation ACA,ACG T380T NP_001166141.1
NM_024706.4 1451 Silent Mutation ACA,ACG T380T NP_078982.3

View Full Product Details