Product Details

SNP ID
rs145458572
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2820960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCACTATCGTCCGCACAGCACTG[C/T]CATCTCCCCACACCCTCCAGGAAGT
Phenotype
MIM: 608159
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRSS21 PubMed Links

Gene Details

Gene
PRSS21
Gene Name
protease, serine 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270452.1 656 Intron NP_001257381.1
NM_006799.3 656 Missense Mutation CCA,TCA P186S NP_006790.1
NM_144956.2 656 Missense Mutation CCA,TCA P184S NP_659205.1
NM_144957.2 656 Missense Mutation CCA,TCA P186S NP_659206.1
XM_017022875.1 656 Missense Mutation CCA,TCA P184S XP_016878364.1
XM_017022876.1 656 Intron XP_016878365.1
XM_017022877.1 656 Intron XP_016878366.1
XM_017022878.1 656 Intron XP_016878367.1
Gene
ZG16B
Gene Name
zymogen granule protein 16B
There are no transcripts associated with this gene.

View Full Product Details