Product Details
- SNP ID
-
rs145594574
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:2586849 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACTGGACTTACAGTTTTCCGAAGA[C/T]GAGAAGAGGTTGTTGTTGGAGAAGC
- Phenotype
-
MIM: 605213
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PDPK1
PubMed Links
Gene Details
- Gene
- PDPK1
- Gene Name
- 3-phosphoinositide dependent protein kinase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001261816.1 |
1010 |
Silent Mutation |
GAC,GAT |
D433D |
NP_001248745.1 |
NM_002613.4 |
1010 |
Silent Mutation |
GAC,GAT |
D433D |
NP_002604.1 |
NM_031268.5 |
1010 |
Silent Mutation |
GAC,GAT |
D306D |
NP_112558.2 |
XM_011522521.2 |
1010 |
Silent Mutation |
GAC,GAT |
D467D |
XP_011520823.1 |
XM_011522523.2 |
1010 |
Silent Mutation |
GAC,GAT |
D340D |
XP_011520825.1 |
XM_017023271.1 |
1010 |
Silent Mutation |
GAC,GAT |
D467D |
XP_016878760.1 |
XM_017023272.1 |
1010 |
Silent Mutation |
GAC,GAT |
D435D |
XP_016878761.1 |
XM_017023273.1 |
1010 |
Silent Mutation |
GAC,GAT |
D433D |
XP_016878762.1 |
XM_017023274.1 |
1010 |
Silent Mutation |
GAC,GAT |
D406D |
XP_016878763.1 |
XM_017023275.1 |
1010 |
Silent Mutation |
GAC,GAT |
D340D |
XP_016878764.1 |
XM_017023276.1 |
1010 |
Silent Mutation |
GAC,GAT |
D306D |
XP_016878765.1 |
XM_017023277.1 |
1010 |
Silent Mutation |
GAC,GAT |
D217D |
XP_016878766.1 |
View Full Product Details