Product Details

SNP ID
rs145844597
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2771585 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTGTTTCTGCTCTTGGCCATCG[C/T]CTGGGAGTGGACATGCAGGCTATGG
Phenotype
MIM: 606032 MIM: 600787
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SRRM2 PubMed Links

Gene Details

Gene
SRRM2
Gene Name
serine/arginine repetitive matrix 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016333.3 819 Intron NP_057417.3
XM_005255226.1 819 Intron XP_005255283.1
XM_006720874.3 819 Intron XP_006720937.1
Gene
TCEB2
Gene Name
transcription elongation factor B subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007108.3 819 UTR 3 NP_009039.1
NM_207013.2 819 Missense Mutation ACG,GCG T125A NP_996896.1

View Full Product Details