Product Details

SNP ID
rs146581011
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:50075687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCCTGTCAGTTCCATGGAATCT[C/T]TTCTATTGAAGACATTGGTAGCAGG
Phenotype
MIM: 614951
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HEATR3 PubMed Links

Gene Details

Gene
HEATR3
Gene Name
HEAT repeat containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005256013.2 659 Missense Mutation CTT,TTT L161F XP_005256070.1
XM_011523188.2 659 Missense Mutation CTT,TTT L130F XP_011521490.1
XM_017023385.1 659 Missense Mutation CTT,TTT L16F XP_016878874.1
XM_017023386.1 659 Missense Mutation CTT,TTT L16F XP_016878875.1

View Full Product Details