Product Details

SNP ID
rs147536970
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56659137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGGGCTGTGTTTGCAAAGGGGC[A/G]TCAGAGAAGTGCAGCTGCTGCGACT
Phenotype
MIM: 156349 MIM: 156352 MIM: 156353
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MT1B PubMed Links

Gene Details

Gene
MT1B
Gene Name
metallothionein 1B
There are no transcripts associated with this gene.

Gene
MT1F
Gene Name
metallothionein 1F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301272.1 275 UTR 3 NP_001288201.1
NM_005949.3 275 Silent Mutation GCA,GCG A53A NP_005940.1
Gene
MT1G
Gene Name
metallothionein 1G
There are no transcripts associated with this gene.

View Full Product Details