Product Details

SNP ID
rs147537253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:71536800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGTCTATGAAGGCACAGCCCGA[A/G]CGCATGCACGTGCTGGTTCTGTCTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHST4 PubMed Links

Gene Details

Gene
CHST4
Gene Name
carbohydrate sulfotransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166395.1 334 Silent Mutation GAA,GAG E41E NP_001159867.1
NM_005769.2 334 Silent Mutation GAA,GAG E41E NP_005760.1

View Full Product Details