Product Details

SNP ID
rs147853124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2712774 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGATCACCCCCGCCTGCAGCCAC[A/G]ACTGACCCACGAGGCACACCAGGGG
Phenotype
MIM: 611285 MIM: 608018
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCTD5 PubMed Links

Gene Details

Gene
KCTD5
Gene Name
potassium channel tetramerization domain containing 5
There are no transcripts associated with this gene.

Gene
PRSS27
Gene Name
protease, serine 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318395.1 839 Missense Mutation TCG,TTG S138L NP_001305324.1
NM_031948.4 839 Missense Mutation TCG,TTG S240L NP_114154.1
XM_011522689.2 839 Missense Mutation TCG,TTG S138L XP_011520991.1

View Full Product Details