Product Details

SNP ID
rs148031885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:50320294 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGACCCAAGAGACAGATCATGTT[C/T]GGAGGGGGTCTGGTGCTCAAACGTT
Phenotype
MIM: 600385
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADCY7 PubMed Links

Gene Details

Gene
ADCY7
Gene Name
adenylate cyclase 7
There are no transcripts associated with this gene.

Gene
BRD7
Gene Name
bromodomain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173984.2 1956 Intron NP_001167455.1
NM_013263.4 1956 Intron NP_037395.2
XM_011523046.2 1956 Intron XP_011521348.1
XM_011523047.2 1956 Silent Mutation CCA,CCG P587P XP_011521349.1
XM_011523048.2 1956 Intron XP_011521350.1
XM_011523049.2 1956 Intron XP_011521351.1
XM_011523050.2 1956 Silent Mutation CCA,CCG P369P XP_011521352.1
XM_017023179.1 1956 Intron XP_016878668.1
XM_017023180.1 1956 Intron XP_016878669.1
XM_017023181.1 1956 Silent Mutation CCA,CCG P368P XP_016878670.1

View Full Product Details