Product Details

SNP ID
rs148259622
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2207170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGGAGGGCTGGGCTTGGGCCCT[A/G]CCTCACCACCCCTGCACCCCAGGCA
Phenotype
MIM: 612190 MIM: 172280
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
BRICD5 PubMed Links

Gene Details

Gene
BRICD5
Gene Name
BRICHOS domain containing 5
There are no transcripts associated with this gene.

Gene
MLST8
Gene Name
MTOR associated protein, LST8 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199173.1 Intron NP_001186102.1
NM_001199174.1 Intron NP_001186103.1
NM_001199175.1 Intron NP_001186104.1
NM_022372.4 Intron NP_071767.3
XM_005255475.3 Intron XP_005255532.2
XM_005255478.2 Intron XP_005255535.1
XM_005255479.2 Intron XP_005255536.1
XM_017023548.1 Intron XP_016879037.1
XM_017023549.1 Intron XP_016879038.1
XM_017023550.1 Intron XP_016879039.1
XM_017023551.1 Intron XP_016879040.1
XM_017023552.1 Intron XP_016879041.1
Gene
PGP
Gene Name
phosphoglycolate phosphatase
There are no transcripts associated with this gene.

View Full Product Details