Product Details

SNP ID
rs148375692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31357051 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGTGCCCCCGGAGGCCGTGAAC[A/C]TGTCCCTGGGCCTGTCCCTGGCGTC
Phenotype
MIM: 151510
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ITGAX PubMed Links

Gene Details

Gene
ITGAX
Gene Name
integrin subunit alpha X
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000887.4 389 Missense Mutation ATG,CTG M90L NP_000878.2
NM_001286375.1 389 Missense Mutation ATG,CTG M90L NP_001273304.1
XM_011545852.1 389 Missense Mutation ATG,CTG M90L XP_011544154.1
XM_011545854.1 389 Missense Mutation ATG,CTG M90L XP_011544156.1

View Full Product Details