Product Details

SNP ID
rs148668779
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1486185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGCCCAGCACGAGGGACCCTCC[A/G]GGGGGGATCTCATAGCCCTCCCTGA
Phenotype
MIM: 613442 MIM: 611140
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTX4 PubMed Links

Gene Details

Gene
PTX4
Gene Name
pentraxin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013658.1 1176 Silent Mutation NP_001013680.1
Gene
TELO2
Gene Name
telomere maintenance 2
There are no transcripts associated with this gene.

View Full Product Details