Product Details

SNP ID
rs150039881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:696932 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGGCGGCAAAACCCTGCAGGTT[C/G]ACGAACTCCTTCTCGCCACCAGGCA
Phenotype
MIM: 609082
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FBXL16 PubMed Links

Gene Details

Gene
FBXL16
Gene Name
F-box and leucine rich repeat protein 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153350.3 802 Silent Mutation GTC,GTG V158V NP_699181.2
Gene
WDR24
Gene Name
WD repeat domain 24
There are no transcripts associated with this gene.

View Full Product Details