Product Details

SNP ID
rs150078051
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67657649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCCTGGCTGTGCATCCATCAGA[A/G]AGTGCAAGGCCCAGGCCATGAGCTG
Phenotype
MIM: 609377 MIM: 610859 MIM: 607484
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ACD PubMed Links
Additional Information
For this assay, SNP(s) [rs14920] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ACD
Gene Name
adrenocortical dysplasia homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001082486.1 1929 Missense Mutation TCT,TTT S531F NP_001075955.1
NM_001082487.1 1929 Missense Mutation TCT,TTT S515F NP_001075956.1
NM_022914.2 1929 Missense Mutation TCT,TTT S528F NP_075065.2
XM_005256115.3 1929 Missense Mutation TCT,TTT S502F XP_005256172.1
Gene
C16orf86
Gene Name
chromosome 16 open reading frame 86
There are no transcripts associated with this gene.

Gene
CARMIL2
Gene Name
capping protein regulator and myosin 1 linker 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013838.1 1929 Intron NP_001013860.1
NM_001317026.1 1929 Intron NP_001303955.1
XM_011522874.1 1929 Intron XP_011521176.1
XM_011522875.2 1929 Intron XP_011521177.2
XM_017022953.1 1929 Intron XP_016878442.1
Gene
ENKD1
Gene Name
enkurin domain containing 1
There are no transcripts associated with this gene.

Gene
PARD6A
Gene Name
par-6 family cell polarity regulator alpha
There are no transcripts associated with this gene.

View Full Product Details