Product Details

SNP ID
rs150240090
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67645262 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCCGGCCCATGGCCCAGACCCCC[A/G]ACGGCATCTCCTGTGAGCTCCGAGG
Phenotype
MIM: 610859 MIM: 604167
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CARMIL2 PubMed Links

Gene Details

Gene
CARMIL2
Gene Name
capping protein regulator and myosin 1 linker 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013838.1 259 Missense Mutation AAC,GAC N6D NP_001013860.1
NM_001317026.1 259 Missense Mutation AAC,GAC N6D NP_001303955.1
XM_011522874.1 259 Missense Mutation AAC,GAC N6D XP_011521176.1
XM_011522875.2 259 Missense Mutation AAC,GAC N6D XP_011521177.2
XM_017022953.1 259 Missense Mutation AAC,GAC N6D XP_016878442.1
Gene
CTCF
Gene Name
CCCTC-binding factor
There are no transcripts associated with this gene.

View Full Product Details