Product Details

SNP ID
rs150354698
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70526695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTAACCTTGCAGAGAGCCACTGG[A/C]ATCAGCTTTGCCATTCATGGAAACT
Phenotype
MIM: 606976 MIM: 605592
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
COG4 PubMed Links

Gene Details

Gene
COG4
Gene Name
component of oligomeric golgi complex 4
There are no transcripts associated with this gene.

Gene
SF3B3
Gene Name
splicing factor 3b subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012426.4 250 Silent Mutation GGA,GGC G13G NP_036558.3
Gene
SNORD111B
Gene Name
small nucleolar RNA, C/D box 111B
There are no transcripts associated with this gene.

View Full Product Details