Product Details

SNP ID
rs150574437
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30424481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAGCTCCTCTTGAAGGGCTGCCC[A/G]TTCCCTGGGGGACCAGCCTTGGGGG
Phenotype
MIM: 615840
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
DCTPP1 PubMed Links

Gene Details

Gene
DCTPP1
Gene Name
dCTP pyrophosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024096.1 337 Missense Mutation CGG,TGG R89W NP_077001.1
Gene
ZNF771
Gene Name
zinc finger protein 771
There are no transcripts associated with this gene.

View Full Product Details