Product Details

SNP ID
rs150696003
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28981106 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGGGCCGTGTTTCCAGGATAGC[A/G]TCTGACTAGATGAACATGCATCACG
Phenotype
MIM: 602354 MIM: 612583
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LAT PubMed Links

Gene Details

Gene
LAT
Gene Name
linker for activation of T-cells
There are no transcripts associated with this gene.

Gene
SPNS1
Gene Name
sphingolipid transporter 1 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142448.1 Intron NP_001135920.1
NM_001142449.1 Intron NP_001135921.1
NM_001142450.1 Intron NP_001135922.1
NM_001142451.1 Intron NP_001135923.1
NM_032038.2 Intron NP_114427.1
XM_006721096.3 Intron XP_006721159.1
XM_017023757.1 Intron XP_016879246.1
XM_017023758.1 Intron XP_016879247.1
XM_017023759.1 Intron XP_016879248.1

View Full Product Details