Product Details

SNP ID
rs150849224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:20862361 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGGCAGGCCTCAGTGGCAGCCT[C/T]GGCCTTCTTGGTCCCGTTGACGAGG
Phenotype
MIM: 616167
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCUN1D3 PubMed Links

Gene Details

Gene
DCUN1D3
Gene Name
defective in cullin neddylation 1 domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173475.3 438 Missense Mutation AAG,GAG K60E NP_775746.1

View Full Product Details