Product Details

SNP ID
rs151301678
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57698092 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGAAATCACCTTAAAGCAGGAGA[C/T]GCTCAGAGACCTGGAGAAGAAGCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADGRG3 PubMed Links

Gene Details

Gene
ADGRG3
Gene Name
adhesion G protein-coupled receptor G3
There are no transcripts associated with this gene.

Gene
DRC7
Gene Name
dynein regulatory complex subunit 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289162.1 232 Missense Mutation ACG,ATG T48M NP_001276091.1
NM_001289163.1 232 Missense Mutation ACG,ATG T48M NP_001276092.1
NM_032269.5 232 Missense Mutation ACG,ATG T48M NP_115645.4
XM_011523377.1 232 Missense Mutation ACG,ATG T48M XP_011521679.1
XM_011523378.1 232 Missense Mutation ACG,ATG T48M XP_011521680.1

View Full Product Details