Product Details

SNP ID
rs138162394
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:11961976 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTACCCTTCCACAGCAGGCCTGG[C/G]AGCCTGGTTTCCAGACCTCCTCAAC
Phenotype
MIM: 603330 MIM: 194524
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNAH9 PubMed Links

Gene Details

Gene
DNAH9
Gene Name
dynein axonemal heavy chain 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001372.3 12913 Missense Mutation GCA,GGA A4318G NP_001363.2
NM_004662.2 12913 Missense Mutation GCA,GGA A630G NP_004653.2
XM_011523703.1 12913 Missense Mutation GCA,GGA A4333G XP_011522005.1
XM_017024292.1 12913 Missense Mutation GCA,GGA A4294G XP_016879781.1
XM_017024293.1 12913 Missense Mutation GCA,GGA A3627G XP_016879782.1
XM_017024294.1 12913 Intron XP_016879783.1
XM_017024295.1 12913 Intron XP_016879784.1
Gene
ZNF18
Gene Name
zinc finger protein 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303281.1 12913 Intron NP_001290210.1
NM_001303282.1 12913 Intron NP_001290211.1
NM_144680.3 12913 Intron NP_653281.2
XM_011524002.2 12913 Intron XP_011522304.1
XM_017025006.1 12913 Intron XP_016880495.1
XM_017025007.1 12913 Intron XP_016880496.1
XM_017025008.1 12913 Intron XP_016880497.1
XM_017025009.1 12913 Intron XP_016880498.1
XM_017025010.1 12913 Intron XP_016880499.1
XM_017025011.1 12913 Intron XP_016880500.1

View Full Product Details