Product Details

SNP ID
rs138219937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:34962185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGTAACAAGTAGTGTGCTGCCA[A/G]ACGATTTCTTTAGTACTAATCCTCC
Phenotype
MIM: 610730
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCT6B PubMed Links

Gene Details

Gene
CCT6B
Gene Name
chaperonin containing TCP1 subunit 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193529.2 656 Intron NP_001180458.1
NM_001193530.1 656 Intron NP_001180459.1
NM_006584.3 656 Intron NP_006575.2
XM_011524203.2 656 Intron XP_011522505.1
XM_017024023.1 656 Intron XP_016879512.1
XM_017024024.1 656 Intron XP_016879513.1
Gene
ZNF830
Gene Name
zinc finger protein 830
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052857.3 656 Missense Mutation AAC,GAC N207D NP_443089.3

View Full Product Details