Product Details

SNP ID
rs138450019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:60043740 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCTGGTCTCTTTCCTGTGGGCT[A/G]TGGGGTGCTCCAGTGTCATCTCCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HEATR6 PubMed Links

Gene Details

Gene
HEATR6
Gene Name
HEAT repeat containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022070.4 5323 Silent Mutation CAC,CAT H1123H NP_071353.4
XM_005257570.3 5323 Silent Mutation CAC,CAT H955H XP_005257627.1
XM_011525110.2 5323 Silent Mutation CAC,CAT H1087H XP_011523412.1
XM_011525111.2 5323 Silent Mutation CAC,CAT H1076H XP_011523413.1
XM_017024936.1 5323 Silent Mutation CAC,CAT H955H XP_016880425.1
XM_017024937.1 5323 Silent Mutation CAC,CAT H955H XP_016880426.1
XM_017024938.1 5323 Silent Mutation CAC,CAT H662H XP_016880427.1
XM_017024939.1 5323 Silent Mutation CAC,CAT H662H XP_016880428.1
Gene
MIR4737
Gene Name
microRNA 4737
There are no transcripts associated with this gene.

View Full Product Details