Product Details

SNP ID
rs138604224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7559880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCCTGCACCTGGTTCCCATTAA[C/T]GCCACCTCCAAGGGTGAGCACTATT
Phenotype
MIM: 612844 MIM: 602695 MIM: 604472
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SENP3 PubMed Links

Gene Details

Gene
SENP3
Gene Name
SUMO1/sentrin/SMT3 specific peptidase 3
There are no transcripts associated with this gene.

Gene
SENP3-EIF4A1
Gene Name
SENP3-EIF4A1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
TNFSF12
Gene Name
tumor necrosis factor superfamily member 12
There are no transcripts associated with this gene.

Gene
TNFSF12-TNFSF13
Gene Name
TNFSF12-TNFSF13 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172089.3 1120 Silent Mutation AAC,AAT N204N NP_742086.1
Gene
TNFSF13
Gene Name
tumor necrosis factor superfamily member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198622.1 1120 Silent Mutation AAC,AAT N97N NP_001185551.1
NM_001198623.1 1120 Silent Mutation AAC,AAT N96N NP_001185552.1
NM_001198624.1 1120 Silent Mutation AAC,AAT N79N NP_001185553.1
NM_003808.3 1120 Silent Mutation AAC,AAT N124N NP_003799.1
NM_172087.2 1120 Intron NP_742084.1
NM_172088.2 1120 Silent Mutation AAC,AAT N124N NP_742085.1

View Full Product Details