Product Details

SNP ID
rs138748634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3513341 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGTTCACTCCAAGGATGGTCTCC[A/T]TTTCCCCGAAGGGTTCCGGAAGGGT
Phenotype
MIM: 607066
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SPATA22 PubMed Links

Gene Details

Gene
SPATA22
Gene Name
spermatogenesis associated 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170695.1 3271 Intron NP_001164166.1
NM_001170696.1 3271 Intron NP_001164167.1
NM_001170697.1 3271 Intron NP_001164168.1
NM_001170698.1 3271 Intron NP_001164169.1
NM_001170699.1 3271 Intron NP_001164170.1
NM_001321336.1 3271 Intron NP_001308265.1
NM_001321337.1 3271 Intron NP_001308266.1
NM_032598.4 3271 Intron NP_115987.2
Gene
TRPV3
Gene Name
transient receptor potential cation channel subfamily V member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258205.1 3271 UTR 3 NP_001245134.1
NM_145068.3 3271 UTR 3 NP_659505.1

View Full Product Details