Product Details

SNP ID
rs138917401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42160331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAAGGAGGGTCTGGGGTCCAAGC[A/G]GAGCCTGCTGGGTGGCCTGGGGGAC
Phenotype
MIM: 604528 MIM: 604037
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNH4 PubMed Links

Gene Details

Gene
KCNH4
Gene Name
potassium voltage-gated channel subfamily H member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012285.2 3096 Silent Mutation TCC,TCT S921S NP_036417.1
XM_017024400.1 3096 Silent Mutation TCC,TCT S921S XP_016879889.1
XM_017024401.1 3096 Silent Mutation TCC,TCT S872S XP_016879890.1
XM_017024402.1 3096 Silent Mutation TCC,TCT S861S XP_016879891.1
XM_017024403.1 3096 Intron XP_016879892.1
Gene
RAB5C
Gene Name
RAB5C, member RAS oncogene family
There are no transcripts associated with this gene.

View Full Product Details