Product Details
- SNP ID
-
rs139098510
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:81715527 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCATCTACGAGACTGTGCGGGACC[A/G]TGTGGCCAAGGGCAGCCAGGGGCCT
- Phenotype
-
MIM: 602375
MIM: 606794
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MRPL12
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2072700] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MRPL12
- Gene Name
- mitochondrial ribosomal protein L12
There are no transcripts associated with this gene.
- Gene
- SLC25A10
- Gene Name
- solute carrier family 25 member 10
View Full Product Details