Product Details

SNP ID
rs139147681
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82946969 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGACCTCAGGTTGTCCTCAAAGGC[A/G]CCCCCTGTGAGGTCCTGTCAGGAGC
Phenotype
MIM: 615337 MIM: 604649
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
B3GNTL1 PubMed Links
Additional Information
For this assay, SNP(s) [rs7225887] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
B3GNTL1
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009905.2 1034 Silent Mutation GGC,GGT G340G NP_001009905.1
NM_001320742.1 1034 Silent Mutation GGC,GGT G326G NP_001307671.1
NM_001320743.1 1034 Silent Mutation GGC,GGT G162G NP_001307672.1
XM_006722272.3 1034 Silent Mutation GGC,GGT G340G XP_006722335.1
XM_006722274.2 1034 Intron XP_006722337.2
XM_011523532.2 1034 Silent Mutation GGC,GGT G318G XP_011521834.1
XM_011523535.1 1034 Silent Mutation GGC,GGT G229G XP_011521837.1
XM_017024205.1 1034 Silent Mutation GGC,GGT G308G XP_016879694.1
XM_017024206.1 1034 Silent Mutation GGC,GGT G223G XP_016879695.1
XM_017024207.1 1034 Intron XP_016879696.1
Gene
LOC105371944
Gene Name
uncharacterized LOC105371944
There are no transcripts associated with this gene.

Gene
TBCD
Gene Name
tubulin folding cofactor D
There are no transcripts associated with this gene.

View Full Product Details