Product Details

SNP ID
rs139157614
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1781331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGGGTGGTAAGTCCAACAAACA[A/G]GATTTCATCTTCTGGAGCTCCTGGA
Phenotype
MIM: 172860
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SERPINF1 PubMed Links

Gene Details

Gene
SERPINF1
Gene Name
serpin family F member 1
There are no transcripts associated with this gene.

Gene
SMYD4
Gene Name
SET and MYND domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052928.2 2783 Silent Mutation TCC,TCT S790S NP_443160.2
XM_011523632.2 2783 Intron XP_011521934.1
XM_011523633.2 2783 Intron XP_011521935.1
XM_017024121.1 2783 Silent Mutation TCC,TCT S686S XP_016879610.1

View Full Product Details