Product Details

SNP ID
rs139574667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48964321 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCAGCGACTCACTCATTCTTCTT[C/T]CCCTTTTGGGCCAGCAGCCAGTTCA
Phenotype
MIM: 137240
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GIP PubMed Links

Gene Details

Gene
GIP
Gene Name
gastric inhibitory polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004123.2 344 Silent Mutation GGA,GGG G82G NP_004114.1

View Full Product Details