Product Details

SNP ID
rs139778490
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3716712 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGAACAGGATGACCAGAATCAC[A/G]ATCAACACCAGAAGTCCACCAACGC
Phenotype
MIM: 609240 MIM: 604682
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
GSG2 PubMed Links

Gene Details

Gene
GSG2
Gene Name
germ cell associated 2, haspin
There are no transcripts associated with this gene.

Gene
ITGAE
Gene Name
integrin subunit alpha E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002208.4 3850 Silent Mutation ATC,ATT I1140I NP_002199.3
XM_011523823.2 3850 Silent Mutation ATC,ATT I1178I XP_011522125.1
XM_011523825.2 3850 Silent Mutation ATC,ATT I1146I XP_011522127.1
XM_011523827.2 3850 Silent Mutation ATC,ATT I1124I XP_011522129.1
XM_011523828.2 3850 Silent Mutation ATC,ATT I1098I XP_011522130.1
XM_017024586.1 3850 Silent Mutation ATC,ATT I1152I XP_016880075.1
XM_017024587.1 3850 Intron XP_016880076.1

View Full Product Details