Product Details

SNP ID
rs139833239
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81868102 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGACAGAAAAGGCAGAGGCAGGA[C/G]AATACCCAGCCTCCTGGATGGTACT
Phenotype
MIM: 601925 MIM: 176790
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARHGDIA PubMed Links

Gene Details

Gene
ARHGDIA
Gene Name
Rho GDP dissociation inhibitor alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185077.2 1885 UTR 3 NP_001172006.1
NM_001185078.2 1885 UTR 3 NP_001172007.1
NM_001301240.1 1885 UTR 3 NP_001288169.1
NM_001301241.1 1885 UTR 3 NP_001288170.1
NM_001301242.1 1885 UTR 3 NP_001288171.1
NM_001301243.1 1885 UTR 3 NP_001288172.1
NM_004309.5 1885 UTR 3 NP_004300.1
XM_011523574.1 1885 UTR 3 XP_011521876.1
Gene
P4HB
Gene Name
prolyl 4-hydroxylase subunit beta
There are no transcripts associated with this gene.

View Full Product Details