Product Details

SNP ID
rs139908935
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:73242954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGATTCGGTGGCATCACGATGCT[C/T]TTCACAGGATACTTCGTCCTGTGTT
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
C17orf80 PubMed Links
Additional Information
For this assay, SNP(s) [rs1566290] are located under a probe and SNP(s) [rs2270728] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
C17orf80
Gene Name
chromosome 17 open reading frame 80
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100621.2 1815 Silent Mutation CTC,CTT L524L NP_001094091.1
NM_001100622.2 1815 Silent Mutation CTC,CTT L560L NP_001094092.1
NM_001288770.1 1815 Silent Mutation CTC,CTT L524L NP_001275699.1
NM_001288771.1 1815 Silent Mutation CTC,CTT L524L NP_001275700.1
NM_017941.5 1815 Silent Mutation CTC,CTT L560L NP_060411.2
XM_005257487.3 1815 Silent Mutation CTC,CTT L560L XP_005257544.1
XM_006721966.3 1815 Silent Mutation CTC,CTT L524L XP_006722029.1
XM_011524961.1 1815 Silent Mutation CTC,CTT L560L XP_011523263.1
XM_011524962.2 1815 Silent Mutation CTC,CTT L560L XP_011523264.1
XM_017024806.1 1815 Silent Mutation CTC,CTT L560L XP_016880295.1
Gene
CPSF4L
Gene Name
cleavage and polyadenylation specific factor 4 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129885.1 1815 Intron NP_001123357.1
XM_011525115.2 1815 Intron XP_011523417.1

View Full Product Details