Product Details

SNP ID
rs140338364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7934190 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGTGTTCGGGGTCAGCTCCAGA[A/C]CATGCTCCAAACCTCACGTGATACA
Phenotype
MIM: 611425 MIM: 604111 MIM: 610969
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CNTROB PubMed Links

Gene Details

Gene
CNTROB
Gene Name
centrobin, centriole duplication and spindle assembly protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037144.5 1154 Missense Mutation AAC,ACC N108T NP_001032221.1
NM_053051.3 1154 Missense Mutation AAC,ACC N108T NP_444279.2
XM_005256437.2 1154 Missense Mutation AAC,ACC N108T XP_005256494.1
XM_005256438.2 1154 Missense Mutation AAC,ACC N108T XP_005256495.1
XM_005256439.2 1154 Missense Mutation AAC,ACC N108T XP_005256496.1
XM_017024128.1 1154 Missense Mutation AAC,ACC N108T XP_016879617.1
XM_017024129.1 1154 Missense Mutation AAC,ACC N108T XP_016879618.1
XM_017024130.1 1154 Missense Mutation AAC,ACC N108T XP_016879619.1
XM_017024131.1 1154 Missense Mutation AAC,ACC N108T XP_016879620.1
XM_017024132.1 1154 Missense Mutation AAC,ACC N108T XP_016879621.1
XM_017024133.1 1154 Missense Mutation AAC,ACC N108T XP_016879622.1
XM_017024134.1 1154 Missense Mutation AAC,ACC N108T XP_016879623.1
XM_017024135.1 1154 Missense Mutation ACA,CCA T18P XP_016879624.1
XM_017024136.1 1154 Missense Mutation ACA,CCA T18P XP_016879625.1
XM_017024137.1 1154 Missense Mutation ACA,CCA T18P XP_016879626.1
XM_017024138.1 1154 Intron XP_016879627.1
XM_017024139.1 1154 UTR 5 XP_016879628.1
XM_017024140.1 1154 UTR 5 XP_016879629.1
XM_017024141.1 1154 Missense Mutation AAC,ACC N108T XP_016879630.1
XM_017024142.1 1154 UTR 5 XP_016879631.1
XM_017024143.1 1154 UTR 5 XP_016879632.1
XM_017024144.1 1154 UTR 5 XP_016879633.1
Gene
KCNAB3
Gene Name
potassium voltage-gated channel subfamily A regulatory beta subunit 3
There are no transcripts associated with this gene.

Gene
TRAPPC1
Gene Name
trafficking protein particle complex 1
There are no transcripts associated with this gene.

View Full Product Details