Product Details

SNP ID
rs140648470
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:15977313 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCAATAGTTTCCTGGACCCAAG[G/T]AAAAGAACTGACCTACTCGTCGAAA
Phenotype
MIM: 600446 MIM: 614535
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ADORA2B PubMed Links

Gene Details

Gene
ADORA2B
Gene Name
adenosine A2b receptor
There are no transcripts associated with this gene.

Gene
ZSWIM7
Gene Name
zinc finger SWIM-type containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042697.1 1125 UTR 3 NP_001036162.1
NM_001042698.1 1125 UTR 3 NP_001036163.1

View Full Product Details