Product Details

SNP ID
rs141438157
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3663853 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGAGCCGCTTGCGGGCCTGGTC[A/G]TGTGTGACCATGGTCATGTCCCAGC
Phenotype
MIM: 606272 MIM: 602836 MIM: 616484
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTNS PubMed Links

Gene Details

Gene
CTNS
Gene Name
cystinosin, lysosomal cystine transporter
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031681.2 423 Intron NP_001026851.2
NM_004937.2 423 Intron NP_004928.2
XM_005256485.2 423 Intron XP_005256542.1
XM_006721463.2 423 Intron XP_006721526.1
XM_006721464.2 423 Intron XP_006721527.1
XM_011523691.2 423 Intron XP_011521993.1
XM_011523692.2 423 Intron XP_011521994.1
XM_017024254.1 423 Intron XP_016879743.1
XM_017024255.1 423 Intron XP_016879744.1
XM_017024256.1 423 Intron XP_016879745.1
XM_017024257.1 423 Intron XP_016879746.1
XM_017024258.1 423 Intron XP_016879747.1
Gene
EMC6
Gene Name
ER membrane protein complex subunit 6
There are no transcripts associated with this gene.

Gene
P2RX5
Gene Name
purinergic receptor P2X 5
There are no transcripts associated with this gene.

Gene
P2RX5-TAX1BP3
Gene Name
P2RX5-TAX1BP3 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
TAX1BP3
Gene Name
Tax1 binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204698.1 423 Silent Mutation CAC,CAT H64H NP_001191627.1
NM_014604.3 423 Silent Mutation CAC,CAT H90H NP_055419.1

View Full Product Details