Product Details

SNP ID
rs141549471
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40661931 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTCTCACCACAGCATGGAGAGA[C/T]TCAATTTCCACTTGCAGGTGGTGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT222 PubMed Links

Gene Details

Gene
KRT222
Gene Name
keratin 222
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152349.2 275 Silent Mutation GAA,GAG E70E NP_689562.1
Gene
LOC105371775
Gene Name
uncharacterized LOC105371775
There are no transcripts associated with this gene.

View Full Product Details