Product Details

SNP ID
rs142014631
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:37491498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACGCTTGGGGGAGGAGAGGTAGT[C/T]GCTTGGTATTTTTGGCTTTCCACCA
Phenotype
MIM: 606618
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP14 PubMed Links

Gene Details

Gene
DUSP14
Gene Name
dual specificity phosphatase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007026.3 1079 Intron NP_008957.1
XM_005256977.3 1079 UTR 5 XP_005257034.1
XM_011524234.1 1079 Intron XP_011522536.1

View Full Product Details