Product Details

SNP ID
rs142136314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63433025 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAAACAGGGAGAGAACAGTCAC[C/T]AGATCCAAGAAGCACATGATCGGTG
Phenotype
MIM: 600019 MIM: 615047
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYB561 PubMed Links

Gene Details

Gene
CYB561
Gene Name
cytochrome b561
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017916.1 2305 UTR 3 NP_001017916.1
NM_001017917.1 2305 UTR 3 NP_001017917.1
NM_001915.3 2305 UTR 3 NP_001906.3
XM_005257091.1 2305 UTR 3 XP_005257148.1
XM_017024260.1 2305 UTR 3 XP_016879749.1
XM_017024261.1 2305 UTR 3 XP_016879750.1
XM_017024262.1 2305 UTR 3 XP_016879751.1
Gene
TANC2
Gene Name
tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 2
There are no transcripts associated with this gene.

View Full Product Details