Product Details

SNP ID
rs142573585
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43400239 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCACTCATGTGCAAGGCTGCAG[C/T]GCTGTGGACGGTCTGGGCCTGCAGC
Phenotype
MIM: 600732
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL4D PubMed Links

Gene Details

Gene
ARL4D
Gene Name
ADP ribosylation factor like GTPase 4D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001661.3 616 Silent Mutation AGC,AGT S169S NP_001652.2
XM_011524782.2 616 Silent Mutation AGC,AGT S169S XP_011523084.1

View Full Product Details