Product Details

SNP ID
rs143018255
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:45169082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGACTCTGGTGGTTCCCTGCCCC[C/T]GACACCGCGGATGGAGAGCCACTCA
Phenotype
MIM: 615695
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HEXIM2 PubMed Links
Additional Information
For this assay, SNP(s) [rs34207107] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
HEXIM2
Gene Name
hexamethylene bisacetamide inducible 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303436.1 1253 Missense Mutation CCG,CTG P67L NP_001290365.1
NM_001303437.1 1253 Missense Mutation CCG,CTG P45L NP_001290366.1
NM_001303438.1 1253 Missense Mutation CCG,CTG P45L NP_001290367.1
NM_001303439.1 1253 Missense Mutation CCG,CTG P45L NP_001290368.1
NM_001303440.1 1253 Missense Mutation CCG,CTG P45L NP_001290369.1
NM_001303441.1 1253 Missense Mutation CCG,CTG P45L NP_001290370.1
NM_001303442.1 1253 Missense Mutation CCG,CTG P45L NP_001290371.1
NM_001303443.1 1253 Missense Mutation CCG,CTG P45L NP_001290372.1
NM_001303444.1 1253 Missense Mutation CCG,CTG P45L NP_001290373.1
NM_144608.2 1253 Missense Mutation CCG,CTG P45L NP_653209.1
XM_006721687.3 1253 Missense Mutation CCG,CTG P68L XP_006721750.1
XM_011524306.2 1253 Missense Mutation CCG,CTG P45L XP_011522608.1
XM_011524307.2 1253 Missense Mutation CCG,CTG P205L XP_011522609.2
XM_011524308.2 1253 Missense Mutation CCG,CTG P206L XP_011522610.2
XM_017024167.1 1253 Missense Mutation CCG,CTG P45L XP_016879656.1
XM_017024168.1 1253 Missense Mutation CCG,CTG P45L XP_016879657.1
XM_017024169.1 1253 Missense Mutation CCG,CTG P45L XP_016879658.1
XM_017024170.1 1253 Missense Mutation CCG,CTG P45L XP_016879659.1
Gene
LOC105371795
Gene Name
uncharacterized LOC105371795
There are no transcripts associated with this gene.

View Full Product Details