Product Details

SNP ID
rs143084569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8436950 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGAGTTCTGGTTTCTTTTCGTGG[G/T]ATTTTTATACCCTTCTCAGCTACAA
Phenotype
MIM: 607538
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NDEL1 PubMed Links

Gene Details

Gene
NDEL1
Gene Name
nudE neurodevelopment protein 1 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025579.2 Intron NP_001020750.1
NM_030808.4 Intron NP_110435.1
XM_011524013.1 Intron XP_011522315.1
XM_017025183.1 Intron XP_016880672.1
XM_017025184.1 Intron XP_016880673.1
XM_017025185.1 Intron XP_016880674.1
XM_017025186.1 Intron XP_016880675.1
XM_017025187.1 Intron XP_016880676.1
XM_017025188.1 Intron XP_016880677.1

View Full Product Details