Product Details

SNP ID
rs143136523
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82032998 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGAGCCCGGCCTCCTTCGAGAAT[A/G]TTCGTGCCAAGGTAGGGCAAGGCTG
Phenotype
MIM: 608347 MIM: 602050
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCXR PubMed Links

Gene Details

Gene
DCXR
Gene Name
dicarbonyl and L-xylulose reductase
There are no transcripts associated with this gene.

Gene
LRRC45
Gene Name
leucine rich repeat containing 45
There are no transcripts associated with this gene.

Gene
RAC3
Gene Name
ras-related C3 botulinum toxin substrate 3 (rho family, small GTP binding protein Rac3)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316307.1 383 Missense Mutation ATT,GTT I93V NP_001303236.1
NM_005052.2 383 Missense Mutation ATT,GTT I93V NP_005043.1

View Full Product Details