Product Details
- SNP ID
-
rs143531189
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:82748926 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCTCCAGGCGCAGCTGGACCTCAT[C/T]GAGAAGGACTATGCTGACCGAGAGG
- Phenotype
-
MIM: 608425
MIM: 604649
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FN3K
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs573054080] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FN3K
- Gene Name
- fructosamine 3 kinase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022158.3 |
601 |
Silent Mutation |
ATC,ATT |
I180I |
NP_071441.1 |
- Gene
- TBCD
- Gene Name
- tubulin folding cofactor D
There are no transcripts associated with this gene.
View Full Product Details