Product Details

SNP ID
rs144042542
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:57868191 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTTTCCTCTTTGACTTCCTCAT[C/T]TTGGTCTTCTCTGAGAAGGCTCGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CUEDC1 PubMed Links

Gene Details

Gene
CUEDC1
Gene Name
CUE domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271875.1 795 Silent Mutation AAA,AAG K331K NP_001258804.1
NM_001292025.1 795 Silent Mutation AAA,AAG K331K NP_001278954.1
XM_006721900.2 795 Silent Mutation AAA,AAG K331K XP_006721963.1
XM_011524812.2 795 Silent Mutation AAA,AAG K224K XP_011523114.1
XM_017024640.1 795 Silent Mutation AAA,AAG K303K XP_016880129.1
XM_017024641.1 795 Silent Mutation AAA,AAG K249K XP_016880130.1

View Full Product Details