Product Details

SNP ID
rs144080462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:32933268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTATCACCAACCCCCACATTGAG[C/G]CCATTCTGGAGAATGAAGACTGGAT
Phenotype
MIM: 615949
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TMEM98 PubMed Links

Gene Details

Gene
TMEM98
Gene Name
transmembrane protein 98
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033504.1 608 Missense Mutation CCC,GCC P76A NP_001028676.1
NM_001301746.1 608 Missense Mutation CCC,GCC P76A NP_001288675.1
NM_015544.2 608 Missense Mutation CCC,GCC P76A NP_056359.2

View Full Product Details