Product Details
- SNP ID
-
rs144635826
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:58206373 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTTCTGAAGGGAGCTCGATTCCA[C/T]GAAGGCCCTGCAGGGAGGCCAGCCA
- Phenotype
-
MIM: 131399
MIM: 609883
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EPX
PubMed Links
Gene Details
- Gene
- EPX
- Gene Name
- eosinophil peroxidase
There are no transcripts associated with this gene.
- Gene
- MKS1
- Gene Name
- Meckel syndrome, type 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001165927.1 |
1587 |
Missense Mutation |
ATG,GTG |
M490V |
NP_001159399.1 |
NM_001321268.1 |
1587 |
Missense Mutation |
ATG,GTG |
M297V |
NP_001308197.1 |
NM_001321269.1 |
1587 |
Missense Mutation |
CAT,CGT |
H472R |
NP_001308198.1 |
NM_017777.3 |
1587 |
Missense Mutation |
ATG,GTG |
M500V |
NP_060247.2 |
XM_005257485.3 |
1587 |
Missense Mutation |
CAT,CGT |
H329R |
XP_005257542.1 |
XM_006721965.2 |
1587 |
Missense Mutation |
CAT,CGT |
H269R |
XP_006722028.1 |
XM_011524957.2 |
1587 |
Missense Mutation |
CAT,CGT |
H475R |
XP_011523259.1 |
XM_011524958.2 |
1587 |
Missense Mutation |
ATG,GTG |
M503V |
XP_011523260.1 |
XM_011524959.2 |
1587 |
Silent Mutation |
TCA,TCG |
S430S |
XP_011523261.1 |
XM_011524960.2 |
1587 |
Intron |
|
|
XP_011523262.1 |
XM_017024803.1 |
1587 |
Silent Mutation |
TCA,TCG |
S427S |
XP_016880292.1 |
XM_017024804.1 |
1587 |
Intron |
|
|
XP_016880293.1 |
XM_017024805.1 |
1587 |
Missense Mutation |
ATG,GTG |
M357V |
XP_016880294.1 |
View Full Product Details